Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

523534

Sigma-Aldrich

3,4-Diethoxybenzyl alcohol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)2C6H3CH2OH
CAS Number:
Molecular Weight:
196.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

mp

39-43 °C (lit.)

SMILES string

CCOc1ccc(CO)cc1OCC

InChI

1S/C11H16O3/c1-3-13-10-6-5-9(8-12)7-11(10)14-4-2/h5-7,12H,3-4,8H2,1-2H3

InChI key

ZOLYZUOLHFXYQD-UHFFFAOYSA-N

General description

3,4-Diethoxybenzyl alcohol can be prepared from 3,4-diethoxybenzaldehyde.[1][2]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of Some Homologs of Papaverine.
Shepard ER and Noth JF.
Journal of the American Chemical Society, 72(10), 4364-4368 (1950)
Degradation of the diarylpropane lignin model compound 1-(3', 4'-diethoxyphenyl)-1, 3-dihydroxy-2-(4''-methoxyphenyl)-propane and derivatives by the basidiomycete Phanerochaete chrysosporium.
Enoki A and Gold MH.
Archives of Microbiology, 132(2), 123-130 (1982)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service