522791
Beryllium acetylacetonate
97%
Synonym(s):
Be(acac)3
About This Item
Recommended Products
assay
97%
form
liquid
reaction suitability
core: beryllium
bp
270 °C (lit.)
mp
100-104 °C (lit.)
density
1.168 g/mL at 25 °C (lit.)
SMILES string
CC(=O)\C=C(\C)O[Be]O\C(C)=C/C(C)=O
InChI
1S/2C5H8O2.Be/c2*1-4(6)3-5(2)7;/h2*3,6H,1-2H3;/q;;+2/p-2/b2*4-3-;
Inchi Key
BBKXDHBLPBKCFR-FDGPNNRMSA-L
signalword
Danger
Hazard Classifications
Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 1B Inhalation - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Inhalation - STOT SE 3
target_organs
Respiratory system, Skin,Respiratory Tract
Storage Class
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service