Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

522473

Sigma-Aldrich

Manganese(II) formate hydrate

Synonym(s):

Manganese formate dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HCO2)2Mn · xH2O
CAS Number:
Molecular Weight:
144.97 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder

form

powder

mp

129.5 °C (lit.)

SMILES string

O.O=CO[Mn]OC=O

InChI

1S/2CH2O2.Mn.H2O/c2*2-1-3;;/h2*1H,(H,2,3);;1H2/q;;+2;/p-2

InChI key

GFAWMMYRQJGQQZ-UHFFFAOYSA-L

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service