Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

522023

Sigma-Aldrich

Methanol-d4

99.8 atom % D

Synonym(s):

Methyl-d3 alcohol d, Tetradeuteromethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3OD
CAS Number:
Molecular Weight:
36.07
Beilstein/REAXYS Number:
1733278
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:

isotopic purity

99.8 atom % D

Quality Level

assay

≥99% (GC)

form

liquid

expl. lim.

5.5-36.5 % (lit.)

packaging

pk of 10 × 0.6 mL

technique(s)

NMR: suitable

impurities

≤0.250% water
water

refractive index

n20/D 1.326 (lit.)

bp

65.4 °C (lit.)

mp

-99 °C (lit.)

density

0.888 g/mL at 25 °C (lit.)

SMILES string

[2H]OC([2H])([2H])[2H]

InChI

1S/CH4O/c1-2/h2H,1H3/i1D3,2D

InChI key

OKKJLVBELUTLKV-MZCSYVLQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methanol-d4 (CD3OD) is a deuterated NMR solvent useful in NMR-based research and analyses. Absolute infrared absorption intensities of CD3OD have been reported between 8000 and 350cm-1.[1]

application

Methanol-d4 may be used as a solvent to analyze the rate of exchange of methoxyl group in camphor and norcamphor dimethyl ketals.[2]

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Infrared intensities of liquids. XVII. Infrared refractive indices from 8000 to 350 cm-1, absolute integrated absorption intensities, transition moments, and dipole moment derivatives of methan-d3-ol and methanol-d4 at 25?C.
Bertie JE and Zhang SL.
J. Chem. Phys. , 101(10), 8364-8379 (1994)
The Rates of Methoxyl Exchange of Camphor and Norcamphor Dimethyl Ketals in Methanol-d4.
Traylor TG and Perrin CL.
Journal of the American Chemical Society, 88(21), 4934-4942 (1966)
Cheng-Kun Lin et al.
Organic & biomolecular chemistry, 13(7), 2100-2107 (2014-12-20)
A straightforward synthesis of novel, 2-heterocyclyl polyhydroxylated pyrrolidines is described. Stereocontrolled additions of nucleophiles to cyclic nitrones generated the corresponding 2,3-trans adducts, allowing the synthesis of the corresponding pyrrolidines via key intermediates bearing an alkyne and a nitrile oxide. Three
Jon Kapla et al.
Physical chemistry chemical physics : PCCP, 17(34), 22438-22447 (2015-08-08)
The disaccharide trehalose (TRH) strongly affects the physical properties of lipid bilayers. We investigate interactions between lipid membranes formed by 1,2-dimyristoyl-sn-glycero-3-phosphocholine (DMPC) and TRH using NMR spectroscopy and molecular dynamics (MD) computer simulations. We compare dipolar couplings derived from DMPC/TRH
William L Scott et al.
Journal of combinatorial chemistry, 11(1), 34-43 (2008-12-25)
For the successful implementation of Distributed Drug Discovery (D(3)) (outlined in the accompanying Perspective), students, in the course of their educational laboratories, must be able to reproducibly make new, high quality, molecules with potential for biological activity. This article reports

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service