Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

520594

Sigma-Aldrich

2-Methyl-5-nitroindole

97%

Synonym(s):

NSC 131898

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8N2O2
CAS Number:
Molecular Weight:
176.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

172-176 °C (lit.)

SMILES string

Cc1cc2cc(ccc2[nH]1)[N+]([O-])=O

InChI

1S/C9H8N2O2/c1-6-4-7-5-8(11(12)13)2-3-9(7)10-6/h2-5,10H,1H3

InChI key

IDJGRXQMAHESOD-UHFFFAOYSA-N

Application

Reactant for preparation of:β
  • Cyanoindoles
  • Methanoindoles
  • Protein kinase C theta (PKCθ) inhibitors
  • Tubulin polymerization inhibitors
  • Peptide-mimetic protease-activated receptor-1 (PAR-1) antagonists
  • Cytosolic phospholipase A2α
  • Cyclooxygenase (COX) inhibitors
  • Serotonin 5-HT6 Receptor Ligands
  • Heterocyclic β-substituted alanine derivatives

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nitration of Indoles. III. Polynitration of 2-Alkylindoles1.
Noland WE, et al.
The Journal of Organic Chemistry, 30(10), 3457-3469 (1965)
Nitration of Indoles. II. The Mononitration of Methylindoles1.
Noland WE, et al.
The Journal of Organic Chemistry, 28(9), 2262-2266 (1963)
Syntheses Based on 2-Methyl-5-nitrogramine. Preparation of 2-Methyl-5-nitroindole-3-acetic Acid.
Noland WE and Rush KR.
The Journal of Organic Chemistry, 28(10), 2921-2922 (1963)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service