Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

520403

Sigma-Aldrich

Copper-zinc alloy

powder, −60 mesh

Synonym(s):

Brass

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
31152304
PubChem Substance ID:
NACRES:
NA.23

form

powder

composition

copper, 70 wt. %
zinc, 30 wt. %

particle size

−60 mesh

SMILES string

[Cu].[Zn]

InChI

1S/Cu.Zn

InChI key

TVZPLCNGKSPOJA-UHFFFAOYSA-N

pictograms

Environment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service