Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

519227

Sigma-Aldrich

4-Ethynylphenylacetonitrile

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC≡CC6H4CH2CN
Molecular Weight:
141.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

refractive index

n20/D 1.5630 (lit.)

density

1.037 g/mL at 25 °C (lit.)

functional group

nitrile

SMILES string

C#Cc1ccc(CC#N)cc1

InChI

1S/C10H7N/c1-2-9-3-5-10(6-4-9)7-8-11/h1,3-6H,7H2

InChI key

CZUBOLAETAAHLA-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

>230.0 °F - closed cup

flash_point_c

> 110 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aggregation and chain dynamics in supramolecular polymers by dynamic rheology: cluster formation and self-aggregation.
Herbst F, et al.
Macromolecules, 43(23), 10006-10016 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service