Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

518468

Sigma-Aldrich

3-Chloropropyl chloroformate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCO2(CH2)3Cl
CAS Number:
Molecular Weight:
157.00
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor pressure

0.15 mmHg ( 20 °C)

assay

97%

refractive index

n20/D 1.45 (lit.)

bp

184-189 °C (lit.)

density

1.302 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

ClCCCOC(Cl)=O

InChI

1S/C4H6Cl2O2/c5-2-1-3-8-4(6)7/h1-3H2

InChI key

MTXMEFUEBCFWCY-UHFFFAOYSA-N

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 1

flash_point_f

205.0 °F - closed cup

flash_point_c

96.1 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Malcolm J D'Souza et al.
International journal of molecular sciences, 21(12) (2020-06-25)
A previous study of the effect of a 2-chloro substituent on the rates and the mechanisms of the solvolysis of ethyl chloroformate is extended to the effect of a 3-chloro substituent on the previously studied solvolysis of propyl chloroformate and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service