Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

516929

Sigma-Aldrich

2-Amino-3-bromo-5-nitrobenzonitrile

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H2(Br)(NO2)CN
CAS Number:
Molecular Weight:
242.03
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

mp

180-185 °C (lit.)

SMILES string

Nc1c(Br)cc(cc1C#N)[N+]([O-])=O

InChI

1S/C7H4BrN3O2/c8-6-2-5(11(12)13)1-4(3-9)7(6)10/h1-2H,10H2

InChI key

MUHLVSZIVTURCZ-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vilas Venunath Patil et al.
The Journal of organic chemistry, 80(16), 7876-7883 (2015-07-28)
Unusual regio- and chemoselective oxidation of aromatic amines hindered with ortho substituents (except -NH2, -NHCH3, and -OH) to the corresponding nitro compounds is described by use of nonanebis(peroxoic acid). The mechanistic investigation for selective oxidation of amines ortho-substituted with -NH2
P David Josephy et al.
Environmental and molecular mutagenesis, 57(1), 10-16 (2015-09-24)
Genotoxicity data on commercial azo dyes and their components remain sparse, despite their widespread use. We have tested the mutagenicity of 2-cyano-4-nitroaniline (CNNA) and 2,6-dicyano-4-nitroaniline (CNCNNA), components of azo dyes such as Disperse Blue 165 and Disperse Red 73, in
The conversion of 2-(4-chloro-5H-1, 2, 3-dithiazolylideneamino) benzonitriles into 3-aminoindole-2-carbonitriles using triphenylphosphine.
Michaelidou SS and Koutentis PA.
Tetrahedron, 65(40), 8428-8433 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service