Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

516759

Sigma-Aldrich

2,5-Dibromobenzoic acid

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br2C6H3CO2H
CAS Number:
Molecular Weight:
279.91
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Quality Level

assay

96%

mp

156-159 °C (lit.)

application(s)

food and beverages

SMILES string

OC(=O)c1cc(Br)ccc1Br

InChI

1S/C7H4Br2O2/c8-4-1-2-6(9)5(3-4)7(10)11/h1-3H,(H,10,11)

InChI key

SQQKOTVDGCJJKI-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Swamping Catalyst Effect. V. The Halogenation of Aromatic Acid Derivatives1.
Pearson DE, et al.
The Journal of Organic Chemistry, 28(11), 3147-3149 (1963)
Selective halogen-lithium exchange in 2, 5-dibromobenzenes and 2, 5-dibromopyridine.
Parham WE and Piccirilli RM.
The Journal of Organic Chemistry, 42(2), 257-260 (1977)
Studies on electrochemical hydrodebromination mechanism of 2, 5-dibromobenzoic acid on Ag electrode by in situ FTIR spectroscopy.
Li MC, et al.
Electrochimica Acta, 56(11), 4100-4104 (2011)
Synthesis of functional polyphenylenes from substituted hydroquinones via nickel (0)-catalyzed polymerization of their bismesylates.
Percec V, et al.
Macromolecules, 28(20), 6726-6734 (1995)
Korey P Carter et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 23(61), 15355-15369 (2017-07-15)
Engaging the nominally terminal oxo atoms of the linear uranyl (UO

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service