Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

516058

Sigma-Aldrich

Methyl (S)-(+)-4,5-dihydro-2-phenyl-4-oxazolecarboxylate

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11NO3
CAS Number:
Molecular Weight:
205.21
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

96%

optical activity

[α]20/D +118°, c = 1 in ethanol

refractive index

n20/D 1.55 (lit.)

bp

131-132 °C/2 mmHg (lit.)

density

1.2 g/mL at 25 °C (lit.)

SMILES string

COC(=O)[C@@H]1COC(=N1)c2ccccc2

InChI

1S/C11H11NO3/c1-14-11(13)9-7-15-10(12-9)8-5-3-2-4-6-8/h2-6,9H,7H2,1H3/t9-/m0/s1

InChI key

DVPGDAXTDJUMAS-VIFPVBQESA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service