515906
Mercury(II) trifluoromethanesulfonate
≥95% trace metals basis
Synonym(s):
Hg(OTf)2
About This Item
Recommended Products
assay
≥95% trace metals basis
reaction suitability
core: mercury
reagent type: catalyst
mp
>350 °C (lit.)
SMILES string
[Hg++].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
InChI
1S/2CHF3O3S.Hg/c2*2-1(3,4)8(5,6)7;/h2*(H,5,6,7);/q;;+2/p-2
InChI key
BPVYMDMPLCOQPJ-UHFFFAOYSA-L
Application
Disclaimer

signalword
Danger
Hazard Classifications
Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2
Storage Class
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service