Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

515701

Sigma-Aldrich

N-(9-Anthracenylmethyl)cinchonindinium chloride

85%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H33ClN2O
CAS Number:
Molecular Weight:
521.09
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

85%

mp

158-161 °C (lit.)

SMILES string

[Cl-].OC(C1CC2CC[N+]1(CC2C=C)Cc3c4ccccc4cc5ccccc35)c6ccnc7ccccc67

InChI

1S/C34H33N2O.ClH/c1-2-23-21-36(22-31-27-11-5-3-9-25(27)19-26-10-4-6-12-28(26)31)18-16-24(23)20-33(36)34(37)30-15-17-35-32-14-8-7-13-29(30)32;/h2-15,17,19,23-24,33-34,37H,1,16,18,20-22H2;1H/q+1;/p-1

InChI key

PICKZKQBJJCSSV-UHFFFAOYSA-M

General description

N-(9-Anthracenylmethyl)cinchonindinium chloride can be prepared by reacting cinchonidine with 9-chloromethylanthracene.[1]

Application

N-(9-Anthracenylmethyl)cinchonindinium chloride may be used in the preparation of O-allyl-N-(9-anthracenylmethyl)cinchonidinium bromide,[1] a cinchona-based chiral phase-transfer catalyst (PTC).[2]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of O-Allyl-N-(9-Anthracenylmethyl) cinchonidinium Bromide as a Phase Transfer Catalyst for the Enantioselective Alkylation of Glycine Benzophenone Imine tert-Butyl Ester:(4S)-2-(Benzhydrylidenamino) pentanedioic Acid, 1-tert-Butyl Ester-5-Methyl Ester.
Corey EJ and Noe MC.
Organic Syntheses, 38-45 (2003)
Cinchona-based phase-transfer catalysts for asymmetric synthesis.
Jew SS and Park HG.
Chemical Communications (Cambridge, England), 46, 7090-7103 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service