Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

51520

Sigma-Aldrich

1,2,3-Benzenetricarboxylic acid hydrate

≥98.0% (calc. based on dry substance, T)

Synonym(s):

Hemimellitic acid hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6O6 · xH2O
CAS Number:
Molecular Weight:
210.14 (anhydrous basis)
Beilstein/REAXYS Number:
2214816
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

≥98.0% (calc. based on dry substance, T)

form

solid

mp

~195 °C (dec.)

functional group

carboxylic acid

SMILES string

OC(=O)c1cccc(C(O)=O)c1C(O)=O

InChI

1S/C9H6O6/c10-7(11)4-2-1-3-5(8(12)13)6(4)9(14)15/h1-3H,(H,10,11)(H,12,13)(H,14,15)

InChI key

UJMDYLWCYJJYMO-UHFFFAOYSA-N

Analysis Note

may contain up to 2 mol water

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystal structure of hemimellitic acid dihydrate (1, 2, 3-benzenetricarboxylic acid dihydrate).
Mo F and Adman E.
Acta Crystallographica Section B, Structural Science, Crystal Engineering and Materials, 31(1), 192-198 (1975)
Derivatives of Hemimellitic Acid. A Synthesis of Erythrocentaurin1.
Wenkert E, et al.
The Journal of Organic Chemistry, 29(9), 2534-2542 (1964)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service