Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

514241

Sigma-Aldrich

2-(tert-Butyldimethylsilyl)-1,3-dithiane

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H22S2Si
CAS Number:
Molecular Weight:
234.50
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

refractive index

n20/D 1.531 (lit.)

bp

124 °C/5 mmHg (lit.)

density

1.01 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)[Si](C)(C)C1SCCCS1

InChI

1S/C10H22S2Si/c1-10(2,3)13(4,5)9-11-7-6-8-12-9/h9H,6-8H2,1-5H3

InChI key

JISLIURJHXAPTN-UHFFFAOYSA-N

Packaging

A versatile acyl anion equivalent.[1][2]

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

221.0 °F - closed cup

flash_point_c

105.00 °C - closed cup

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A B Smith et al.
Organic letters, 1(12), 2001-2004 (2000-06-03)
[formula: see text] An efficient synthesis of trisacetonide (+)-11, the Schreiber C(16-28) subtarget for mycoticins A and B, is described. The key synthetic transformation entails a one-flask, five-component linchpin coupling tactic.
Amos B Smith et al.
The Journal of organic chemistry, 71(7), 2547-2557 (2006-03-25)
N-Toluenesulfonyl aziridines comprise effective second electrophiles in the solvent controlled three-component linchpin union of silyl dithianes for the stereocontrolled convergent elaboration of protected 1,5-amino alcohols. This tactic, in conjunction with a one-flask sequential cyclization, constitutes an effective general strategy for
Amos B Smith et al.
Organic letters, 7(15), 3247-3250 (2005-07-16)
[reaction: see text]. A stereocontrolled total synthesis of the neotropical poison-frog alkaloid (-)-205B (1) has been achieved, employing a dithiane three-component linchpin coupling, a one-pot sequential construction of the embedded indolizidine ring, and ring-closing metathesis (RCM) to arrive at the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service