Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

514152

Supelco

Chlorine dioxide release mixture

Aseptrol® WTS 7.05 sachet

Synonym(s):

Aseptrol® WTS 7.05 sachet, Sodium chlorite

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

manufacturer/tradename

Aseptrol® WTS 7.05 sachet

mp

190 °C (dec.)

SMILES string

[Na+].[O-]Cl=O

InChI

1S/ClHO2.Na/c2-1-3;/h(H,2,3);/q;+1/p-1

Inchi Key

UKLNMMHNWFDKNT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Aseptrol is a registered trademark of Engelhard Corp.

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Chronic 2 - Eye Dam. 1 - Ox. Sol. 1 - Skin Corr. 1B - STOT RE 2

target_organs

spleen

supp_hazards

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service