Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

513229

Sigma-Aldrich

4-Methylbenzhydrylamine hydrochloride, polymer-bound

80-150 mesh, extent of labeling: 0.5-1.0 mmol/g N loading, 1 % cross-linked with divinylbenzene

Synonym(s):

(α-Amino-α-p-xylyl hydrochloride)polystyrene crosslinked with divinylbenzene, MBHA resin, MBHA-polystyrene

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352116
NACRES:
NA.22

crosslinking

1 % cross-linked with divinylbenzene

extent of labeling

0.5-1.0 mmol/g N loading

particle size

80-150 mesh

application(s)

peptide synthesis

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service