Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

512540

Sigma-Aldrich

Bis(pentamethylcyclopentadienyl)magnesium

electronic grade, 99.999% trace metals basis

Synonym(s):

MgCp2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H30Mg
CAS Number:
Molecular Weight:
294.76
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

electronic grade

assay

99.999% trace metals basis

form

solid

reaction suitability

core: magnesium

mp

230 °C (dec.) (lit.)

SMILES string

[Mg].C[C]1[C](C)[C](C)[C](C)[C]1C.C[C]2[C](C)[C](C)[C](C)[C]2C

InChI

1S/2C10H15.Mg/c2*1-6-7(2)9(4)10(5)8(6)3;/h2*1-5H3;

InChI key

LIEOGMBLMOOSAY-UHFFFAOYSA-N

General description

Atomic number of base material: 12 Magnesium

pictograms

Flame

signalword

Danger

hcodes

Hazard Classifications

Pyr. Sol. 1

Storage Class

4.2 - Pyrophoric and self-heating hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service