Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

512427

Sigma-Aldrich

Dibutyl butylphosphonate

≥90%

Synonym(s):

DBBP, Dibutoxybutylphosphine oxide, NSC 2666

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3P(O)[O(CH2)3CH3]2
CAS Number:
Molecular Weight:
250.31
Beilstein/REAXYS Number:
1777999
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

assay

≥90%

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.432 (lit.)

bp

283 °C (lit.)

density

0.946 g/mL at 25 °C (lit.)

SMILES string

CCCCOP(=O)(CCCC)OCCCC

InChI

1S/C12H27O3P/c1-4-7-10-14-16(13,12-9-6-3)15-11-8-5-2/h4-12H2,1-3H3

InChI key

JPGXOMADPRULAC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chemical used for:
  • Uranium extraction from concentrated phosphoric acid[1]
  • Plasticizing[2][3][4]

Other Notes

Remainder tributyl phosphate

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H. M. Abu Shawish, et al.,
Materials Science and Engineering, C, 31, 300-306 (2011)
Vinod Kumar Gupta et al.
Talanta, 85(2), 970-974 (2011-07-06)
Poly(vinyl chloride) (PVC)-based membranes of N,N-bis-succinamide-based dendritic molecule with tetrabutyl ammonium bromide (TBAB) as a cation inhibitor and dibutylphthalate (DBP), dioctylphthalate (DOP), dibutyl (butyl) phosphonate (DBBP) and 1-chloronaphthalene (CN) as plasticizing solvent mediators were prepared and used as nitrate ion-selective
B. Sethi, et al.,
Journal of Electroanalytical Chemistry (Lausanne Switzerland), 651, 185-190 (2011)
A. A. Abdel-Khalek, et al.,
J. Radioanal. Nucl. Chem., 290, 353-359 (2011)
Ma de Lourdes Ballinas et al.
Environmental science & technology, 38(3), 886-891 (2004-02-19)
Polymer inclusion membranes (PIMs) based on cellulose triacetate (CTA) and dibutyl butyl phosphonate (DBBP) were tested for arsenic(V) separation from H2SO4 for its recovery from copper electrolytes. Solvent extraction experiments allowed the determination of the As(V)-DBBP and H2SO4-DBBP complexes formed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service