Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

511587

Sigma-Aldrich

Perruthenate, polymer-supported

16-50 mesh, extent of labeling: 4.5-4.8 mmol N per g loading

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352128
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

reaction type: solution phase peptide synthesis
reactivity: alcohol reactive

extent of labeling

4.5-4.8 mmol N per g loading

particle size

16-50 mesh

SMILES string

[O-][Ru](=O)(=O)=O.C=Cc1ccccc1.C=Cc2ccc(C=C)cc2.C[N+](C)(C)Cc3ccc(C=C)cc3

Application

This resin is used to oxidize primary and secondary alcohols to aldehydes and ketone, respectively and to oxidize secondary hydroxylamines to provide nitrones.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hinzen, B.
Synthesis, 977-977 (1998)
Hinzen, B,; Ley, S.V.
Journal of the American Chemical Society, 1907-1907 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service