511463
Poly(acrylamide-co-acrylic acid) partial sodium salt
average Mw 200,000, acrylamide ~20 wt. %
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
mol wt
average Mw 200,000
composition
acrylamide, ~20 wt. %
impurities
<7 wt. % water
solubility
water and some protic solvents: soluble
SMILES string
[Na+].NC(=O)C=C.OC(=O)C=C.[O-]C(=O)C=C
InChI
1S/C3H5NO.2C3H4O2.Na/c3*1-2-3(4)5;/h2H,1H2,(H2,4,5);2*2H,1H2,(H,4,5);/q;;;+1/p-1
InChI key
DTBMFUOWAASNDB-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Application
Binder for cellulose fibers, iron oxide to tape, pigments to TV tubes. Thickener and stabilizer, flocculant and protective colloid.
Features and Benefits
Insolubilized by trivalent metal salts, notably alum.
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service