Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

510602

Sigma-Aldrich

6-Phenyl-3(2H)-pyridazinone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8N2O
CAS Number:
Molecular Weight:
172.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

mp

201-204 °C (lit.)

SMILES string

O=C1NN=C(C=C1)c2ccccc2

InChI

1S/C10H8N2O/c13-10-7-6-9(11-12-10)8-4-2-1-3-5-8/h1-7H,(H,12,13)

InChI key

IJUIPRDMWWBTTQ-UHFFFAOYSA-N

Gene Information

human ... PTPRA(5786)

General description

6-Phenyl-3(2H)-pyridazinone is a pyridazinone derivative that can be prepared from 4,5- dihydro-6-phenyl-3(2H)-pyridazinone.[1] It is reported to be a weak positive inotropic agent.[2] 6-Phenyl-3(2H)-pyridazinone forms the core of compounds that show a potential antiplatelet activity.[1]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cardiotonic agents. 2. Synthesis and structure-activity relationships of 4,5-dihydro-6-[4-(H-imidazol-1-yl)phenyl]-3(2H)-pyridazinones: a new class of positive inotropic agents.
Sircar I, et al.
Journal of Medicinal Chemistry, 28(10), 1405-1413 (1985)
Eddy Sotelo et al.
Bioorganic & medicinal chemistry, 10(9), 2873-2882 (2002-07-12)
A series of 6-phenyl-3(2H)-pyridazinones with a diverse range of substituents in the 5-position have been prepared and evaluated in the search for new antiplatelet agents. A significant dependence of the substituent on the inhibitory effect has been observed. The pharmacological

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service