Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

50598

Sigma-Aldrich

2-Nitrophenol polymer bound

extent of labeling: ~1.3 mmol/g loading

Synonym(s):

(4-Hydroxy-3-nitrobenzamidomethyl)polystyrene

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
PubChem Substance ID:

reaction suitability

reaction type: solution phase peptide synthesis

extent of labeling

~1.3 mmol/g loading

matrix

crosslinked with 2% DVB

particle size

200-400 mesh

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jae Wook Lee et al.
Journal of combinatorial chemistry, 5(3), 330-335 (2003-05-13)
Novel nitrophenol solid supports based on various resin materials (polystyrene, TentaGel, macroporous, PEGA, and silica gel) are reported for facile amide and sulfonamide library synthesis. The broad choice of resin materials available will allow the reaction to occur successfully in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service