All Photos(1)
About This Item
Linear Formula:
BrC6H4ZnI
CAS Number:
Molecular Weight:
348.29
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
concentration
0.5 M in THF
density
1.045 g/mL at 25 °C
storage temp.
2-8°C
SMILES string
Brc1ccc([Zn]I)cc1
InChI
1S/C6H4Br.HI.Zn/c7-6-4-2-1-3-5-6;;/h2-5H;1H;/q;;+1/p-1
InChI key
LQPWHJBKOBCQRS-UHFFFAOYSA-M
Legal Information
Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
target_organs
Respiratory system
supp_hazards
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
1.0 °F - closed cup
flash_point_c
-17.22 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service