Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

497509

Sigma-Aldrich

Tetrakis(dimethylsilyl) orthosilicate

96%

Synonym(s):

Tetrakis(dimethylsiloxy)silane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[HSi(CH3)2O]4Si
CAS Number:
Molecular Weight:
328.73
Beilstein/REAXYS Number:
1784368
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Quality Level

assay

96%

refractive index

n20/D 1.387 (lit.)

bp

190 °C (lit.)

density

0.884 g/mL at 25 °C (lit.)

SMILES string

C[SiH](C)O[Si](O[SiH](C)C)(O[SiH](C)C)O[SiH](C)C

InChI

1S/C8H28O4Si5/c1-13(2)9-17(10-14(3)4,11-15(5)6)12-16(7)8/h13-16H,1-8H3

InChI key

GTWPLTBXXGOHNT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

152.6 °F - closed cup

flash_point_c

67 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Winston H Elliott et al.
Journal of biomechanical engineering (2019-09-17)
Time-dependent arterial wall property is an important but difficult topic in vascular mechanics. Hysteresis, which appears during the measurement of arterial pressure-diameter relationship through a cardiac cycle, has been used to indicate time-dependent mechanics of arteries. However, the cause-effect relationship

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service