Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

497444

Sigma-Aldrich

Dichloro[(R,R)-ethylenebis(4,5,6,7-tetrahydro-1-indenyl)]titanium(IV)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24Cl2Ti
CAS Number:
Molecular Weight:
383.18
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

optical activity

[α]21/D +3489°, c = 0.07 in methylene chloride

mp

175-180 °C (lit.)

SMILES string

Cl[Ti]1(Cl)[C@]4(CC[C@]12C=CC3=C2CCCC3)C=CC5=C4CCCC5

InChI

1S/C20H24.2ClH.Ti/c1-3-7-19-15(5-1)9-11-17(19)13-14-18-12-10-16-6-2-4-8-20(16)18;;;/h9-12H,1-8,13-14H2;2*1H;/q;;;+2/p-2

InChI key

JCEDTJKHTKLRJD-UHFFFAOYSA-L

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

supp_hazards

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service