About This Item
Recommended Products
assay
95%
refractive index
n20/D 1.462 (lit.)
bp
133-135 °C/10 mmHg (lit.)
density
1.03 g/mL at 25 °C (lit.)
functional group
chloro
ether
SMILES string
ClCCCCCCOC1CCCCO1
InChI
1S/C11H21ClO2/c12-8-4-1-2-5-9-13-11-7-3-6-10-14-11/h11H,1-10H2
InChI key
XOLOZSPDGPUSFV-UHFFFAOYSA-N
General description
Application
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service