Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

497096

Sigma-Aldrich

Poly(4-hydroxy benzoic acid-co-ethylene terephthalate)

80 mol % in 4-hydroxybenzoic acid

Synonym(s):

1,4-benzenedicarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(OC6H4CO)x(OCH2CH2O2CC6H4-4-CO)y
CAS Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

liquid crystal

impact strength

225 J/m (Izod)

concentration

80 mol % in 4-hydroxybenzoic acid

SMILES string

OC(=O)c1ccc(O)cc1.OCCOC(=O)c2ccc(cc2)C(O)=O

Looking for similar products? Visit Product Comparison Guide

Physical form

Liquid crystalline copolyester

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service