Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

497088

Sigma-Aldrich

Poly(ethylenecarbonyl-co-propylenecarbonyl)

Synonym(s):

Alternating carbon monoxide-ethylene-propylene copolymer, Carbon monoxide-ethene-propene copolymer, Carbon monoxide-ethylene-propylene alternating copolymer, Carbon monoxide-ethylene-propylene copolymer, Carbon monoxide-ethylene-propylene terpolymer, Ethene-propene-carbon monoxide copolymer, Ethylene-propylene-carbon monoxide copolymer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH2CH2CO)x(CH2CH2CH2CO)y
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23
Pricing and availability is not currently available.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service