495174
Barium bis(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionate)
99.99% trace metals basis
Synonym(s):
Ba(FOD)2
About This Item
assay
99.99% trace metals basis
form
solid
reaction suitability
core: barium
mp
92-96 °C (lit.)
SMILES string
CC(C)(C)C(\O[Ba]O\C(=C/C(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)C(C)(C)C)=C\C(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
InChI
1S/2C10H11F7O2.Ba/c2*1-7(2,3)5(18)4-6(19)8(11,12)9(13,14)10(15,16)17;/h2*4,18H,1-3H3;/q;;+2/p-2/b2*5-4-;
InChI key
NCPWXEUADOWGIO-SJGYQHGCSA-L
General description
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Faceshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service