495158
Calcium bis(6,6,7,7,8,8,8,-heptafluoro-2,2-dimethyl-3,5-octanedionate)
≥99.9% trace metals basis
Synonym(s):
Ca(FOD)2
About This Item
assay
≥99.9% trace metals basis
reaction suitability
core: calcium
mp
189.1-193.5 °C (lit.)
SMILES string
CC(C)(C)\C(O[Ca]O\C(=C/C(=O)C(F)(F)C(F)(F)C(F)(F)F)C(C)(C)C)=C\C(=O)C(F)(F)C(F)(F)C(F)(F)F
InChI
1S/2C10H11F7O2.Ca/c2*1-7(2,3)5(18)4-6(19)8(11,12)9(13,14)10(15,16)17;/h2*4,18H,1-3H3;/q;;+2/p-2/b2*5-4-;
InChI key
UUXRFZXYYNXEBG-SJGYQHGCSA-L
General description
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service