Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

494798

Sigma-Aldrich

Trichloroacetimidate on Wang resin

200-400 mesh, extent of labeling: 0.75-1.25 mmol/g trichloroacetimidate loading, 1 % cross-linked

Synonym(s):

4-Alkoxybenzyl trichloroacetimidate, polymer-bound, Wang trichloroacetimidate resin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

crosslinking

1 % cross-linked

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

extent of labeling

0.75-1.25 mmol/g trichloroacetimidate loading

particle size

200-400 mesh

SMILES string

C=Cc1ccccc1.C=Cc2ccc(C=C)cc2.ClC(Cl)(Cl)C(=N)OCc3ccc(OCc4ccccc4)cc3

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service