Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

49463

Sigma-Aldrich

(2R,4S,5R)-2-Hydroxymethyl-5-ethylquinuclidine

≥98.0% (GC)

Synonym(s):

(2R,4S,5R)-5-Ethyl-2-quinuclidinylmethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H19NO
CAS Number:
Molecular Weight:
169.26
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

≥98.0% (GC)

optical activity

[α]/D +178±5°, c = 1 in chloroform

technique(s)

gas chromatography (GC): suitable

storage temp.

2-8°C

SMILES string

CC[C@H]1CN2CC[C@H]1C[C@@H]2CO

InChI

1S/C10H19NO/c1-2-8-6-11-4-3-9(8)5-10(11)7-12/h8-10,12H,2-7H2,1H3/t8-,9-,10+/m0/s1

InChI key

CWZWBZXPEJJRSS-LPEHRKFASA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service