Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

494585

Sigma-Aldrich

Lithium diisopropylamide solution

1.8 M in THF/heptane/ethylbenzene

Synonym(s):

LDA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CH]2NLi
CAS Number:
Molecular Weight:
107.12
Beilstein/REAXYS Number:
3655042
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

concentration

1.8 M in THF/heptane/ethylbenzene

bp

65 °C

density

0.836 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

[Li]N(C(C)C)C(C)C

InChI

1S/C6H14N.Li/c1-5(2)7-6(3)4;/h5-6H,1-4H3;/q-1;+1

InChI key

ZCSHNCUQKCANBX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Titanium-ate enolates prepared from ketones by Ti(OiPr)4 and LDA underwent efficient radical trifluoromethylation providing trifluoromethyl ketones in good yield.[1] Also used in directed ortholithiation of arylsulfonyloxazolidinones to prepare N-substituted saccharin analogs.[2]

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT RE 2 - STOT SE 3

target_organs

Central nervous system, hearing organs, Respiratory system

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

10.4 °F - closed cup

flash_point_c

-12 °C - closed cup

ppe

Faceshields, Gloves, Goggles


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 47, 6405-6405 (2006)
Paweł Mandera et al.
Quarterly journal of experimental psychology (2006), 68(8), 1623-1642 (2015-02-20)
Subjective ratings for age of acquisition, concreteness, affective valence, and many other variables are an important element of psycholinguistic research. However, even for well-studied languages, ratings usually cover just a small part of the vocabulary. A possible solution involves using
Erwei Yin et al.
IEEE transactions on neural systems and rehabilitation engineering : a publication of the IEEE Engineering in Medicine and Biology Society, 23(4), 693-701 (2015-02-24)
The present study proposes a hybrid brain-computer interface (BCI) with 64 selectable items based on the fusion of P300 and steady-state visually evoked potential (SSVEP) brain signals. With this approach, row/column (RC) P300 and two-step SSVEP paradigms were integrated to
Putri W Novianti et al.
BMC bioinformatics, 16, 199-199 (2015-06-22)
Class prediction models have been shown to have varying performances in clinical gene expression datasets. Previous evaluation studies, mostly done in the field of cancer, showed that the accuracy of class prediction models differs from dataset to dataset and depends
Journal of Fluorine Chemistry, 127, 539-539 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service