Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

494194

Sigma-Aldrich

(S)-(+)-1-(2-Chlorophenyl)-1,2-ethanediol

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CH(OH)CH2OH
CAS Number:
Molecular Weight:
172.61
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

assay

96%

optical activity

[α]20/D +70°, c = 0.2 in chloroform

bp

293 °C (lit.)

mp

68-75 °C (lit.)

SMILES string

OC[C@@H](O)c1ccccc1Cl

InChI

1S/C8H9ClO2/c9-7-4-2-1-3-6(7)8(11)5-10/h1-4,8,10-11H,5H2/t8-/m1/s1

InChI key

YGOPULMDEZVJGI-MRVPVSSYSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

226.4 °F - closed cup

flash_point_c

108 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service