Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

487317

Sigma-Aldrich

2-Fluoro-5-methylphenyl isocyanate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(CH3)NCO
CAS Number:
Molecular Weight:
151.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

refractive index

n20/D 1.511 (lit.)

bp

192 °C (lit.)

density

1.159 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

Cc1ccc(F)c(c1)N=C=O

InChI

1S/C8H6FNO/c1-6-2-3-7(9)8(4-6)10-5-11/h2-4H,1H3

InChI key

GLSUJZPVKMKUPJ-UHFFFAOYSA-N

General description

2-Fluoro-5-methylphenyl isocyanate, also known as 1-fluoro-2-isocyanato-4-methylbenzene, is an organic building block containing an isocyanate group. Its enthalpy of vaporization at boiling point has been reported.[1]

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

185.0 °F - closed cup

flash_point_c

85 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaws CL.
Thermophysical Properties of Chemicals and Hydrocarbons, 549-549 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service