Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

483125

Sigma-Aldrich

2-Methyl-1-hexen-3-yne

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C≡CC(CH3)=CH2
CAS Number:
Molecular Weight:
94.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

refractive index

n20/D 1.45 (lit.)

bp

130 °C (lit.)

density

0.758 g/mL at 25 °C (lit.)

SMILES string

CCC#CC(C)=C

InChI

1S/C7H10/c1-4-5-6-7(2)3/h2,4H2,1,3H3

InChI key

IXPWKHNDQICVPZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Flame

signalword

Danger

hcodes

pcodes

Hazard Classifications

Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

39.2 °F - closed cup

flash_point_c

4 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karima Ait-Aissa et al.
Arteriosclerosis, thrombosis, and vascular biology, 42(9), 1121-1136 (2022-07-29)
Radiation therapy strongly increases the risk of atherosclerotic vascular disease, such as carotid stenosis. Radiation induces DNA damage, in particular in mitochondria, but the upstream and downstream signaling events are poorly understood. The objective of this study was to define
Weiwei Yi et al.
Cell reports, 34(13), 108922-108922 (2021-04-01)
Hematopoietic stem cells (HSCs) regenerate blood cells upon hematopoietic injuries. During homeostasis, HSCs are maintained in a low reactive oxygen species (ROS) state to prevent exhaustion. However, the role of nitric oxide (NO) in controlling HSC regeneration is still unclear.
Alessandra Sacchi et al.
Cell death & disease, 11(10), 921-921 (2020-10-29)
The immunological mechanisms underlying the clinical presentation of SARS-CoV-2 infection and those influencing the disease outcome remain to be defined. Myeloid-derived suppressor cells (MDSC) have been described to be highly increased during COVID-19, however, their role remains elusive. We performed
Chunxiu Zhou et al.
Antioxidants (Basel, Switzerland), 11(7) (2022-07-28)
3,4',5-trimethoxy-trans-stilbene (TMS) is a methoxylated derivative of resveratrol. Previous studies showed the vaso-protective effects of resveratrol; nevertheless, research on this derivative is scarce. The current study aimed to explore whether TMS can alleviate endothelial dysfunction in diabetic and obese mice

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service