Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

483079

Sigma-Aldrich

Potassium tungstate

≥99.99% trace metals basis

Synonym(s):

Dipotassium tetraoxotungstate, Potassium tungstate (K2 WO4 )

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2WO4
CAS Number:
Molecular Weight:
326.03
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

assay

≥99.99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: tungsten

mp

921 °C (lit.)

density

3.12 g/mL at 25 °C (lit.)

SMILES string

[K+].[K+].[O-][W]([O-])(=O)=O

InChI

1S/2K.4O.W/q2*+1;;;2*-1;

InChI key

AAQNGTNRWPXMPB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service