Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

481572

Sigma-Aldrich

Aldolase Antibody 38C2, murine catalytic monoclonal antibody

average Mw ~150,000, powder (Lyophilized)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200

antibody product type

primary antibodies

form

chunks
powder (Lyophilized)

mol wt

average Mw ~150,000

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ALDOA(226)

Looking for similar products? Visit Product Comparison Guide

Application

Catayzes antibody for aldol reactions; first antibody to be commercially available for organic synthesis applications.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service