Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

481068

Sigma-Aldrich

Poly(carbodihydridosilane)

Synonym(s):

HPCS, Hydridopolycarbosilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH4Si)n
CAS Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

refractive index

n20/D 1.509 (lit.)

bp

50 °C (lit.)

density

0.95 g/mL at 25 °C (lit.)

Storage temp.

2-8°C

SMILES string

[H][Si]([H])=C

Application

Silicon carbide precursor.
Wear coating and sealer coating for ceramic composites and precursor for porous composites for high-temperature filters and catalysts.

Physical properties

Chlorine-free

Pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

78.8 °F - closed cup

flash_point_c

26 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service