Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

480681

Sigma-Aldrich

6-O-(tert-Butyldiphenylsilyl)-D-galactal cyclic carbonate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H26O5Si
CAS Number:
Molecular Weight:
410.54
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

assay

97%

optical activity

[α]20/D −40.0°, c = 1 in chloroform

contains

5% ethyl acetate as mobility inducer

mp

−3 °C (lit.)

storage temp.

−20°C

SMILES string

CC(C)(C)[Si](OC[C@@H](O)[C@@H]1OC(=O)O[C@H](C=O)[C@H]1O)(c2ccccc2)c3ccccc3

InChI

1S/C23H28O7Si/c1-23(2,3)31(16-10-6-4-7-11-16,17-12-8-5-9-13-17)28-15-18(25)21-20(26)19(14-24)29-22(27)30-21/h4-14,18-21,25-26H,15H2,1-3H3/t18-,19-,20-,21+/m1/s1

InChI key

OXQFQHKHZPNZSN-NCYKPQTJSA-N

Application

Important building block for both solution- and solid-phase synthesis of oligosaccharides.[1]

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seeberger, P.H., et al.
Aldrichimica Acta, 30, 75-75 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service