Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

480207

Sigma-Aldrich

6-(Dibutylamino)-1,8-diazabicyclo[5.4.0]undec-7-ene

98%

Synonym(s):

6-Dibutylamino-1,8-diazabicyclo[5.4.0]undec-7-ene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H33N3
CAS Number:
Molecular Weight:
279.46
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

assay

98%

refractive index

n20/D 1.5 (lit.)

bp

312 °C (lit.)

solubility

H2O: insoluble
organic solvents: soluble

density

0.942 g/mL at 25 °C (lit.)

SMILES string

CCCCN(CCCC)C1CCCCN2CCCN=C12

InChI

1S/C17H33N3/c1-3-5-12-19(13-6-4-2)16-10-7-8-14-20-15-9-11-18-17(16)20/h16H,3-15H2,1-2H3

InChI key

MGKYTFDYDXZTEM-UHFFFAOYSA-N

Application

Accelerator for polymerization reactions.

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service