Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47992

Sigma-Aldrich

(+)-β-Funebrene

≥97.0% (sum of enantiomers, GC)

Synonym(s):

(+)-1,7-Diepi-β-cedrene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24
CAS Number:
Molecular Weight:
204.35
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

assay

≥97.0% (sum of enantiomers, GC)

optical activity

[α]20/D +9±1°, neat

refractive index

n20/D 1.503

density

0.933 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

C[C@@H]1CC[C@H]2C(C)(C)[C@@H]3C[C@]12CCC3=C

InChI

1S/C15H24/c1-10-7-8-15-9-12(10)14(3,4)13(15)6-5-11(15)2/h11-13H,1,5-9H2,2-4H3/t11-,12-,13+,15-/m1/s1

InChI key

DYLPEFGBWGEFBB-GUIRCDHDSA-N

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service