Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

479012

Sigma-Aldrich

Ethyl 3-isocyanatopropionate

98%

Sign Into View Organizational & Contract Pricing

Select a Size

2 G
$107.35
10 G
$172.50

$107.35

List Price$113.00Save 5%
Web-Only Promotion

Available to ship onApril 29, 2025Details


Request a Bulk Order

Select a Size

Change View
2 G
$107.35
10 G
$172.50

About This Item

Linear Formula:
OCNCH2CH2CO2C2H5
CAS Number:
Molecular Weight:
143.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$107.35

List Price$113.00Save 5%
Web-Only Promotion

Available to ship onApril 29, 2025Details


Request a Bulk Order

Quality Level

assay

98%

refractive index

n20/D 1.429 (lit.)

bp

68 °C/3 mmHg (lit.)

density

1.087 g/mL at 25 °C (lit.)

functional group

amine
ester
isocyanate

storage temp.

2-8°C

SMILES string

CCOC(=O)CCN=C=O

InChI

1S/C6H9NO3/c1-2-10-6(9)3-4-7-5-8/h2-4H2,1H3

InChI key

XBSGYVHOINMTIM-UHFFFAOYSA-N

General description

Ethyl 3-isocyanatopropionate, also known as ethyl N-(oxomethylene)-β-alaninate, is an aliphatic isocyanate.

Application

Ethyl 3-isocyanatopropionate may be used in the synthesis of ethyl N-{[(3-[(tert-butoxycarbonyl)(-cholestan-3-yl)amino]propyl)amino]carbonyl}-β-alaninate.[1]

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

199.4 °F - closed cup

flash_point_c

93 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Siwarutt Boonyarattanakalin et al.
Journal of the American Chemical Society, 128(35), 11463-11470 (2006-08-31)
Binding of ligands to macromolecular receptors on the surface of mammalian cells often results in ligand uptake through receptor-mediated endocytosis. Certain human leukocytes and epithelial cells express Fc receptors (FcRs) that bind and internalize antibodies through this mechanism. To mimic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service