Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

478466

Sigma-Aldrich

Undecyl isocyanate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)10NCO
CAS Number:
Molecular Weight:
197.32
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

refractive index

n20/D 1.44 (lit.)

bp

103 °C/3 mmHg (lit.)

density

0.868 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCN=C=O

InChI

1S/C12H23NO/c1-2-3-4-5-6-7-8-9-10-11-13-12-14/h2-11H2,1H3

InChI key

JXAYHHMVMJVFPQ-UHFFFAOYSA-N

General description

Undecyl isocyanate (n-undecylisocyanate) is an alkyl isocyanate that can be synthesized by reacting lauroyl chloride with sodium azide.

Application

Undecyl isocyanate may be used in the synthesis of poly(n-undecyl isocyanate) (PUDIC) and starch-N-undecyl carbamate.

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of poly (n-undecyl isocyanate)/polyaniline polyblend.
Li YJ, et al.
Journal of Applied Polymer Science, 117(1), 1-7 (2010)
Preparation of starch carbamates in homogeneous phase using different mixing conditions.
Engelmann G, et al.
Starch/Staerke, 53(11), 560-569 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service