Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

478423

Sigma-Aldrich

Phenyl isocyanatoformate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
OCNCO2C6H5
CAS Number:
Molecular Weight:
163.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

refractive index

n20/D 1.516 (lit.)

bp

100 °C/15 mmHg (lit.)

density

1.233 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

O=C=NC(=O)Oc1ccccc1

InChI

1S/C8H5NO3/c10-6-9-8(11)12-7-4-2-1-3-5-7/h1-5H

InChI key

FSZKLYCUEQGCKW-UHFFFAOYSA-N

General description

Phenyl isocyanatoformate, also known as phenyl carbonisocyanatidate, is an organic building block containing an isocyanate group. It participates in the preparation of resin-bound 1,7-disubstituted-1,3,5-triazepane-2,4-dione.[1]

Application

Phenyl isocyanatoformate may be used in the synthesis of 8-(β-D-2′-deoxy-3′,5′-bis(4-toluoyloxy)ribofuranosyl)-imidazo[1,2-a]-1,3,5-triazine-2(8H),4(3H)-dione[2] and 1,3,5-benzotriazepine-2,4-diones.[3]

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

210.2 °F - closed cup

flash_point_c

99 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of a 1, 3, 5-benzotriazepine-2,4-dione based library.
Chuckowree I, et al.
Tetrahedron Letters, 53(28), 3607-3611 (2012)
Y Yu et al.
Organic letters, 3(18), 2797-2799 (2001-09-01)
[reaction: see text]. The solid-phase synthesis of 1,7-disubstituted-1,3,5-triazepane-2,4-diones from resin-bound amino acids is described. The exhaustive reduction of solid-support bound amides with borane afforded the requisite secondary amines, which following treatment with phenyl isocyanatoformate and cleavage, provided the corresponding triazepane-2,4-diones.
P Rao et al.
The Journal of organic chemistry, 66(15), 5012-5015 (2001-07-21)
A concise route is described to prepare the 5-aza-7-deazapurine 2'-deoxyriboside (4), which presents the puADA hydrogen-bonding pattern, analogous to the hydrogen-bonding pattern presented by 2'-deoxyxanthosine (2). The route begins with the commercially available 1-alpha-chloro-2-deoxy-3-5-bistoluoyloxyribofuranose (10), which proves to be a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service