Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

478369

Sigma-Aldrich

2-Chloro-4-nitrophenyl isocyanate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(NO2)NCO
CAS Number:
Molecular Weight:
198.56
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Product 478369 is not currently sold in your country. Contact Technical Service

assay

99%

form

solid

mp

59-63 °C (lit.)

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1ccc(N=C=O)c(Cl)c1

InChI

1S/C7H3ClN2O3/c8-6-3-5(10(12)13)1-2-7(6)9-4-11/h1-3H

InChI key

POLDSAUWRJCSOK-UHFFFAOYSA-N

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service