Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

477885

Sigma-Aldrich

3-Benzyloxybenzaldehyde, polymer-bound

200-400 mesh, extent of labeling: 1.5-2.0 mmol/g loading, 1 % cross-linked

Synonym(s):

3-Benzyloxyaldehyde, polymer-bound

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

crosslinking

1 % cross-linked

extent of labeling

1.5-2.0 mmol/g loading

particle size

200-400 mesh

Application

Photolabile safety-catch linker intermediate.
Polystyrene-based resin for SPOS.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 38, 1227-1227 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service