477826
N-(Phosphonomethyl)iminodiacetic acid hydrate
95%
About This Item
Recommended Products
assay
95%
form
solid
mp
215 °C (dec.) (lit.)
SMILES string
[H]O[H].OC(=O)CN(CC(O)=O)CP(O)(O)=O
InChI
1S/C5H10NO7P.H2O/c7-4(8)1-6(2-5(9)10)3-14(11,12)13;/h1-3H2,(H,7,8)(H,9,10)(H2,11,12,13);1H2
InChI key
QTDRAPLUBFHWCT-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service