Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47655

Sigma-Aldrich

Bis(phenylthio)methane

≥98.0% (GC)

Synonym(s):

Diphenylthiomethane, Formaldehyde diphenyl mercaptal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2(SC6H5)2
CAS Number:
Molecular Weight:
232.36
Beilstein/REAXYS Number:
1874146
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Quality Level

assay

≥98.0% (GC)

form

solid

bp

194 °C/8 mmHg (lit.)

mp

34-37 °C (lit.)

SMILES string

C(Sc1ccccc1)Sc2ccccc2

InChI

1S/C13H12S2/c1-3-7-12(8-4-1)14-11-15-13-9-5-2-6-10-13/h1-10H,11H2

InChI key

ZHUPZVIALZHGGP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bis(phenylthio)methane (PhSCH2SPh) is a dithioether ligand. It forms coordination complex with silver(I) salts.[1][2][3] It participates in the synthesis of (-)-rasfonin.[4]

Application

Bis(phenylthio)methane (PhSCH2SPh) may be used in the synthesis of the following:
  • dihydroxy thioethers[5]
  • [Cu4I4{μ-PhS2CH2SPh}2]n[6]
  • [(η5-C5Me5)MCl21-(PhS)2CH2)] (M=Rh, Ir ; C5Me5= pentamethylcyclopentadienyl)[7]
  • 6-deoxy-1,2:3,4-di-O-isopropylidene-α-D-galactoheptodialdo-1,5-pyranose diphenyl dithioacetal[8]

Other Notes

Acyl anion equivalent with some advantages over 1,3-dithiane;[9][10] for the preparation of vinyl sulfides.[11]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

6-Deoxy-1,2: 3,4-di-O-isopropylidene-α-d-galacto-heptodialdo-1,5-pyranose diphenyl dithioacetal.
Weaver TD, et al.
Acta Crystallographica Section E, Structure Reports Online, 60(11), 2113-2114 (2004)
Methane-derived polyanionic synthons from bis (phenylthio) methane.
Foubelo F, et al.
Tetrahedron Letters, 40(46), 8177-8180 (1999)
A concise asymmetric synthesis of (-)-rasfonin.
Huang Y, et al.
Organic & Biomolecular Chemistry, 10(1), 29-31 (2012)
Valderrama M, et al.
Journal of Organometallic Chemistry, 620(1), 256-262 (2001)
Awaleh MO, et al.
Crystal Growth & Design, 5(5), 1897-1906 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service